Learn More
Vector Laboratories Sambucus Nigra Lectin CY3 1 mg L-1010-10
SDP
Supplier:  Vector Laboratories L101010
Encompass
Sambucus Nigra Lectin (SNA, EBL), CY3, 1 mg, isolated from elderberry bark, binds preferentially to sialic acid attached to terminal galactose in alpha-2,6 and, to a lesser degree, alpha-2,3 linkage. Binding is also inhibited to some extent by lactose or galactose. This lectin does not appear to bind sialic acid linked to N-acetylgalactosamine. SNA has been reported to inhibit cell-free protein synthesis. Cy3-labeled Sambucus nigra lectin has an appropriate number of fluorochromes bound to provide the optimum staining characteristics for this lectin. This conjugate is supplied essentially free of unconjugated fluorochromes. Excitation: 550 nm; Emission: 570 nm; Color: Red.
Catalog No. 50-204-5664