Learn More
Medchemexpress LLC SBP-0636457 | 1422180-49-1 | 98.4% | 456.58 | C25H36N4O4 | 25MG

Supplier: Medchemexpress LLC HY12537825MG
SBP-0636457 is a SMAC mimetic that antagonizes inhibitor of apoptosis proteins (IAPs) and is used in preclinical research to study apoptosis and cancer cell viability. The compound is provided as a solid with reported analytical purity and recommended storage conditions for stability.
- SMAC mimetic and IAP antagonist for apoptosis research.
- High reported purity suitable for analytical and cellular assays.
- Off-white to yellow solid form for convenient handling and formulation.
- Molecular weight and formula provided for experimental calculations.
- Recommended storage conditions specified to maintain compound stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.