Learn More
Selleck Chemical LLC SBP-3264-E1065-5MG
SDP
Supplier:  Selleck Chemical LLC E10655MG
Encompass_Preferred
SBP-3264 is a valuable chemical probe for understanding the roles of STK3 and STK4 in acute myeloid leukemia (AML)
Catalog No. 50-313-7216