Learn More
Cayman Chemical SBromoenol lactoned7
SDP
Supplier:  Cayman Chemical 105351 MG
Encompass_Preferred
An internal standard for the quantification of (S)-BEL by GC- or LC-MS
Catalog No. 50-288-7662