Learn More
TARGETMOL CHEMICALS INC SC-26196 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1285610MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. SC-26196 is an orally active inhibitor of Delta6 desaturase (D6D) with IC50 of 0.2 uM in a rat liver microsomal assay with antiinflammatory properties. purity: 99%
Catalog No. 50-258-1412