Learn More
Apexbio Technology LLC SC 26196 218136-59-5 10mg
SDP
Supplier:  Apexbio Technology LLC B760810
Encompass_Preferred
SC 26196 is a small-molecule inhibitor targeting Delta6-desaturase (D6D FADS2) It is designed to inhibit D6D an enzyme catalyzing the desaturation of linoleic and -linolenic acids into longer-chain polyunsaturated fatty acids thereby modulating fatty acid metabolic pathways SC 26196 exerts its biological activity primarily through inhibition of D6D In rat liver microsome assays SC 26196 demonstrates inhibitory activity with an IC50 value of approximately 0 2 M Based on these pharmacological properties SC 26196 holds research potential in the regulation of lipid metabolism and the study of inflammatory processes in metabolic and inflammation-related disease models
Catalog No. 50-000-87846