Learn More
TARGETMOL CHEMICALS INC SC-43 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T84785MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SC-43 is a potent and orally active agonist of SHP-1 (PTPN6). SC-43 inhibits the phosphorylation of STAT3 and induces cell apoptosis and with anti-fibrotic and anticancer effects. purity: 99%
Catalog No. 50-257-9069