Learn More
Selleck Chemical LLC SC-514 S4907-10mg
SDP
Supplier:  Selleck Chemical LLC S490710MG
Encompass_Preferred
SC-514 (GK 01140) is an orally active ATP-competitive IKK-2 inhibitor with IC50 of 3-12 M blocks NF- B-dependent gene expression does not inhibit other IKK isoforms or other serine-threonine and tyrosine kinases
Catalog No. 50-311-2120