Learn More
Selleck Chemical LLC SC-560
SDP
Supplier:  Selleck Chemical LLC S668625MG
Encompass_Preferred
SC-560 is a highly selective COX-1 inhibitor with IC50 of 9 nM compared with the IC50 of 6 3 M for COX-2 nearly 1 000-fold higher than with COX-1
Catalog No. 50-000-02384