Learn More
Apexbio Technology LLC SC 560 188817-13-2 5mg
SDP
Supplier:  Apexbio Technology LLC B67865
Encompass_Preferred
SC 560 (CAS 188817-13-2) is a selective inhibitor of cyclooxygenase-1 (COX-1) exhibiting an IC50 of 0 009 M for COX-1 and 6 3 M for COX-2 By preferentially blocking COX-1 activity SC 560 impedes the conversion of arachidonic acid to prostaglandins such as thromboxane and PGE2 In vitro studies demonstrate that SC 560 suppresses COX-1-derived prostaglandin generation in a concentration-dependent manner inhibits cellular proliferation and induces apoptosis In vivo administration results in complete inhibition of COX-1-mediated thromboxane B2 and PGE2 production and attenuation of tumor growth in ovarian epithelial cancer models SC 560 serves as a valuable tool for investigating COX-1 s roles in inflammation pain and tumorigenesis
Catalog No. 50-000-88446