Learn More
TARGETMOL CHEMICALS INC SC-560 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T410125MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. SC-560 is a member of the diaryl heterocycle class of cyclooxygenase (COX) inhibitors. Purity 99.39%
Catalog No. 50-287-7970