Learn More
Enzo Life Sciences SC-560 (5mg). CAS: 188817-13-2
SDP
Supplier:  Enzo Life Sciences ALX270427M005
Encompass_Preferred
Selective COX-1 inhibitor (IC50=9nM). Shows 700-fold more selectivity over COX-2 (IC50=6.3µM). Inhibits COX-1-derived platelet thromboxane B2, gastric PGE2, and dermal PGE2 production. Orally active in rat. Alternative name: 5-(4-chlorophenyl)-1-(4-methoxyphenyl)-3-(trifluormethyl)-1H-pyrazole. Purity: ≥98%. Appearance: White to off-white crystalline solid. Solubility: Soluble in 100% ethanol, DMSO or dimethyl formamide; sparingly soluble in aqueous buffers. Long Term Storage: -20°C.
Catalog No. 50-200-8550