Learn More
Selleck Chemical LLC SC-58125-E0789-5MG
SDP
Supplier:  Selleck Chemical LLC E07895MG
Encompass_Preferred
SC-58125 is a potent and selective inhibitor of human cyclooxygenase 2 (hCOX-2) and triple mutant of hCOX-2 with an IC50 of 0 04 M and 1 M respectively
Catalog No. 50-313-7389