Learn More
TARGETMOL CHEMICALS INC SC-75416 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T287031MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. SC-75416 is a selective COX-2 inhibitor for the study of pain. Purity 98.67%
Catalog No. 50-287-9306