Learn More
Apexbio Technology LLC SC 79 305834-79-1 50mg
SDP
Supplier:  Apexbio Technology LLC B566350
Encompass_Preferred
SC 79 (CAS 305834-79-1) is a small-molecule activator of the serine/threonine kinase Akt (protein kinase B) a primary downstream effector in the phosphatidylinositol-3-phosphate signaling cascade known for its anti-apoptotic functions Identified through chemical genetic screening SC 79 binds specifically to Akt s PH domain promoting its activation in the cytosolic compartment independently of membrane translocation This activation facilitates Akt phosphorylation by upstream kinases enhancing cell survival Research indicates SC 79 confers protective effects in cellular assays and animal models of ischemic stroke suggesting potential therapeutic relevance across neurological research applications
Catalog No. 50-000-63905