Learn More
Apexbio Technology LLC SC-9 102649-78-5 10mg
SDP
Supplier:  Apexbio Technology LLC B630210
Encompass_Preferred
SC-9 (CAS 102649-78-5) is a small molecule modulator that functions as an activator of protein kinase C (PKC) a family of enzymes involved in regulating intracellular signaling pathways By enhancing PKC enzymatic activity SC-9 influences cellular processes such as differentiation proliferation and survival This compound is frequently utilized in biomedical research to investigate the roles of PKC in various physiological and pathological contexts supporting studies aimed at elucidating disease mechanisms and informing the development of novel therapeutic approaches
Catalog No. 50-000-90856