Learn More
Strem, An Ascensus Company 5g. Tris(N,N'-di-i-propylformamidinato)scandium(III), (99.9%-Sc).
SDP
Supplier:  Strem, An Ascensus Company 2112005G
Encompass
5g. Tris(N,N'-di-i-propylformamidinato)scandium(III), (99.9%-Sc). . Molecular Weight: 426.58. Molecular Formula: C21H45ScN6. Color/form: white to off-white pwdr. Strem# 21-1200. http://www.strem.com/catalog/v/21-1200/
Catalog No. 50-204-5271