Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec Pump Tubing, 2-Stop, Tygon S3™ E-Lab, 1.14 mm ID; 12/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9646030/DEL
Encompass
Appearance: Opaque Temperature: -70 to 52¿C Life at 100 rpm: 1000 hrs
The overall length of E-Lab (E-3603) 96460-series stop tubing is 16" (406 mm). The legacy Ismatec stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
Catalog No. NC1666743