Learn More
Hach Company SC1000 Disp w/Modbus TCP/GSM
Supplier:  Hach Company LXV402.99.12002
Encompass
SC1000 Multi-parameter Universal Controller Display Module with Modbus TCP/IP and GSM/GPRS protocols (for use in USA) Includes display module with communication (as appropriate)-basic user module
Catalog No. 50-002-07801
Add to Cart
Request a Quote