Learn More
Hach Company SC4500 Ctrl Claros 5x mA Out 1
Supplier:  Hach Company LXV525.99EA1541
Encompass
SC4500 Controller-Claros-enabled-5x mA Output-1 digital Sensor-1 mA Input-100-240 VAC-US plugIncludes SC4500 Controller-Claros-enabled-with EU plug includes mounting hardware
Catalog No. 50-002-07343