Learn More
Hach Company SC4500 Ctrl Prog 5x mA Out 2 a
Supplier:  Hach Company LXV525.99P11111
Encompass
SC4500 Controller-Prognosys-5x mA Output-2 analog pH/ORP Sensors-without plug C1D2Includes SC4500 Controller-Non-Claros Embedded Prognosys-without plug C1D2 includes mounting hardware
Catalog No. 50-002-07369