Learn More
Cayman Chemical SC514 Inhibitors
SDP
Supplier:  Cayman Chemical 1001026710 MG
Encompass_Preferred
A selective and reversible inhibitor of IKK2 (IC50 = 3-12 μM) that displays >10-fold selectivity over 28 other kinases; attenuates NF-κB-mediated gene expression in synovial fibroblasts, smooth muscle cells, and astrocytes
Catalog No. 50-288-8384