Learn More
Aviva Systems Biology Corp SC5DL Antibody - N-terminal region (ARP44751_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44751_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SC5DL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SC5DL. Alternate names: ERG3, S5DES, SC5D, SC5DLApplicationWB. Species reactivity: Human
Catalog No. 50-014-58890