Learn More
Medchemexpress LLC HY-10496 10mg Medchemexpress, SC75741 CAS:913822-46-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1049610MG
Encompass_Preferred
Medchemexpress, HY-10496 10mg SC75741 CAS:913822-46-5 SC75741 is a broad and efficient NF-κB inhibitor with an IC50 of 200 nM for p65. SC75741 blocks influenza viruses (IV) replication in non-toxic concentrations. SC75741 impairs DNA binding of the NF-κB subunit p65, resulting in reduced expression of cytokines, chemokines, and pro-apoptotic factors. SC75741 subsequently inhibits caspase activation and blocks caspase-mediated nuclear export of viral ribonucleoproteins. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8237