Learn More
TARGETMOL CHEMICALS INC SC75741 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T666110MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SC75741 is a potent P65 inhibitor with IC50 of 200 nMpurity: 99%
Catalog No. 50-257-6478