Learn More
Medchemexpress LLC SC79 | 305834-79-1 | 98.0% | 364.78 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY18749200MG
Encompass_Preferred
SC79 is a unique, specific, and BBB permeable Akt activator. It activates Akt in the cytosol and inhibits Akt membrane translocation, specifically binding to the PH domain of Akt. It is for research use only.
  • Augments Akt phosphorylation at both Thr308 and S473 sites.
  • Reduces neuronal excitotoxicity and prevents stroke-induced neuronal death.
  • Suppresses PHAKTM-GFP plasma membrane translocation.
  • Protects against fas-induced fulminant hepatic failure.
  • Protects hepatocytes from TNFα-mediated apoptosis and from Gal/LPS-induced liver injury and damage.
Catalog No. 50-003-88246