Learn More
Essendant Brecknell Portable Electronic Utility Bench Scale, 100 lb Capacity, 12.5 x 10.95 x 2.2 Platform
Supplier:  Essendant SBWGP100
Encompass_Preferred
Portable bench scale which is designed for greater accuracy with attached handle Weight hold key locks weight reading when a larger package blocks the display Internal backlit display with four 1 high LCD digits for easy visibility Discreet keyboard technology Electro-mechanic design 22-gauge steel base Auto-zero tracking and manual zero return Programmable auto-shut-down Scale Type- Utility Freight Electronic/Mechanical- Electronic Increments- 0 1 kg 0 2 lb Power Source(s)- AC Adapter (Sold Separately) 9V Battery (Included)
Catalog No. 50-000-28608