Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Scale Reticle RET 16, 21 mm, 10 mm Scale, 0.05 mm x 200 Divisions
SDP

Supplier:  Electron Microscopy Sciences 6027421

Encompass_Preferred

High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 21 mm
Scale: 10 mm
Divisions: 0.05 mm x 200

Catalog No. 50-190-1751


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.