Learn More
Med Vet International Exel Sterile Disposable Scalpel #15, stainless steel, 10/Box
Supplier:  Med Vet International 29556
Encompass_Preferred
These products do not contain Natural Rubber Latex. Computer controlled grinding, sharpening and polishing, ensures consistently smooth incision propertiesSterilized by GAMMA radiation 2.5 – RADSA Vapor Corrosion Inhibitor (VCI) paper lining, leakage testing, and an aluminum foil peal pack help insure quality and integrity when ready for use. Each production lot is sampled and conforms to ISO 2859, inspection level (S4) AQL 1. & 2.5Blade rigidity is tested to eliminate blade breakageBlades conform to ISO: 774 (1995) & BS: 2982 (1986)Blades are ultrasonically cleaned prior to packing Made from premium quality Swiss carbon steel and British Stainless Steel
Catalog No. 50-264-3626