Learn More
Essendant Epson Expression 13000XL Archival Scanner, Scans Up to 12.2" x 17.2", 4800 dpi Optical Resolution
Supplier:  Essendant EPSB11B257201
Encompass
Take large-format photo and graphics scanning to a new level with the Epson Expression 13000XL Engineered for demanding applications such as photo archiving and graphic arts this powerful scanner uses the latest technology to truly capture your originals a 2400x4800 dpi resolution and optical density of 3 8 Dmax allow for clean sharp images additionally Epson MatrixCCD and ReadyScan LED technologies plus 48-bit color depth deliver brilliant vibrant colors with incredible accuracy The Epson Expression 13000XL brings exceptional versatility to the table accommodating reflective and other media types up to 12 2x17 2 Add on Epson s optional Transparency Unit (sold separately) to easily scan film and other transparent materials up to 12x16 5 One-touch color restoration using Easy Photo Fix technology batch scanning and powerful LaserSoft Imaging SilverFast AI scanning and image processing software help enhance your productivity and expand the limits of what a professional scanne
Catalog No. 50-000-32539