Learn More
Essendant Epson Perfection V850 Pro Scanner, Scans Up to 8.5" x 11.7", 6400 dpi Optical Resolution
Supplier:  Essendant EPSB11B224201
Encompass
The pro-quality scanner with advanced control to archive film and photos Get unparalleled scans with state-of-the-art 6400 dpi optical resolution and advanced optics High-reflection mirror and anti-reflective optical coatings ensure minimum optical distortion Epson s exclusive Dual Lens System automatically selects from two lenses for optimum results based on the media being scanned Digital ICE Technologies remove many surface defects from film and prints to minimize costly retouching ReadyScan LED light source for fast scans with no warm-up time required Batch scanning for multiple slides negatives medium- and large-format film with two sets of film holders (included) Scan Size (WxH)- 8 5x11 7 Color Bit Depth- 48-bit Grayscale Bit Depth- 16-bit Connector/Port/Interface- USB 2 0
Catalog No. 50-000-35775