Learn More
Essendant Epson WorkForce DS-60000 Scanner, 600 dpi Optical Resolution, 200-Sheet Duplex Auto Document Feeder
Supplier:  Essendant EPSB11B204221
Encompass
Capture large two-sided documents in a single scan through the ADF or the scanner bed Achieve remarkable results thanks to the four-line CCD sensor (RGB and black) that provides vivid color and crisp sharp text Boost productivity with one-pass two-sided scanning at speeds of 40 ppm/80 ipm Work more efficiently thanks to the built-in 200-sheet automatic document feeder plus auto duplex scanning Ultrasonic double-feed detector automatically stops the scanning process if two pages feed together Use the optional networking module and included Document Capture Pro software to scan to any PC on the local network Scan directly to popular cloud services like SharePoint Evernote Google Docs and more Scan and save documents to searchable PDF files Includes powerful productivity software Epson Scan Document Capture Pro (Windows only) and ABBYY FineReader Sprint OCR One-touch scanning creates up to 30 user-programmable jobs Scan Size (WxH)- 11 7x17 Color Bit Depth- 16-bit Conn
Catalog No. 50-000-34227