Learn More
Cayman Chemical SCatechol OMethyltrnsfRas
SDP
Supplier:  Cayman Chemical 15563100 UG
Encompass_Preferred
Source: Active recombinant N-terminal hexahistidine- and SUMOpro-tagged COMT expressed in E. coli • Amino acids: 2-221 (full length) • MW: 37.6 kDa
Catalog No. 50-296-9388