Learn More
Cayman Chemical Scavenger Receptor B2 CD36 Mo
SDP
Supplier:  Cayman Chemical 100098935 EA
Encompass_Preferred
Immunogen: Recombinant full-length mouse CD36 expressed in adenovirus • Host: CD36 null mouse • Species Reactivity: (+) Human, mouse, rat • Cross Reactivity: (+) CD36 • Applications: FC, ICC
Catalog No. 50-288-6605