Learn More
Medchemexpress LLC Scavenger receptor class B member 1 (SCARB1) recombinant protein, human | 95.0% | 20 UG
SDP
Supplier:  Medchemexpress LLC HYP7665820UG
Encompass_Preferred
Recombinant human SR-BI/SCARB1 ectodomain (P33-Y443) produced in HEK293 cells and supplied with a His-Fc affinity tag. Supplied as a 20 μg preparation with reported purity of 95.0%, it is suitable for biochemical assays, ligand-binding studies, and antibody development where native glycosylation is preferred.
  • Recombinant human ectodomain P33-Y443 expressed in HEK293 cells.
  • His-Fc affinity tag for capture and detection.
  • Supplied as a 20 μg preparation for small-scale assays.
  • Reported purity 95.0% suitable for biochemical and binding assays.
  • Includes accession identifier Q8WTV0-1/NP_001076428.1 for sequence reference.
Catalog No. 50-002-90409