Learn More
Grainger SCBA Cylinder White
Supplier:  Grainger 16N402
Encompass_Preferred
The Honeywell North SCBA Cylinder is an aluminum air cylinder designed for use with self-contained breathing apparatus (SCBA) systems. It has a capacity of 45 cubic feet and is rated for 4500 psi working pressure. The cylinder features an aluminum liner and carbon fiber composite wrap construction. It has a CGA thread connection and includes a cylinder valve. The cylinder is manufactured in accordance with DOT specifications and is marked with a DOT serial number. It weighs approximately 27 lbs when empty. The cylinder provides high pressure breathing air to SCBA wearers in hazardous environments.
Catalog No. 50-276-3898