Learn More
Abcam SCD1 Inhibitor, SCD1(stearoyl-CoA desaturase 1) inhibitor, 1MG
Supplier:  Abcam AB1420891MG
Encompass_Preferred
MW 387.9 Da, Purity >99%. Potent, selective SCD1 (stearoyl-CoA desaturase 1) inhibitor (IC₅₀ = 4.5 nM). Inhibits the conversion of saturated, long-chain fatty acyl-CoAs to monounsaturated, long-chain fatty acyl-CoAs in vitro, when heptadecanoic acid and palmitic acid are used as the substrate (IC₅₀ = 7.9 and 6.8 nM, respectively). Active in vivo. Orally bioavailable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3329