Learn More
Medchemexpress LLC SCF Rat HEK293 10ug

Supplier: Medchemexpress LLC HYP7107A10UG
SCF Protein, Rat (HEK293) is a recombinant rat stem cell factor produced in HEK293 cells. This hematopoietic cytokine binds the c-Kit receptor and supports hematopoiesis, spermatogenesis, and melanogenesis; it is routinely used for stem/progenitor cell expansion, hematopoietic stem cell mobilization, and related research applications.
- Expression system: HEK293 cells.
- Source species: rat.
- Molecular weight: approximately 10-24 kDa (SDS-PAGE, reducing).
- Purity: >95% by reducing SDS-PAGE.
- Formulation: lyophilized from 0.2 μm filtered buffer (e.g., PBS or 20 mM PB, 150 mM NaCl, pH 7.4).
- Storage: store lyophilized at -20°C; aliquot and store at -20°C/-80°C for extended storage after reconstitution.
- Reconstitution guidance: avoid reconstituting to concentrations below 100 μg/mL in ddH2O.
- Applications: stem/progenitor cell expansion and hematopoietic research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.