Learn More
Medchemexpress LLC Sch-1473759 hydrochloride | 1094067-13-6 | 99.2% | 463.00 | C20H27ClN8OS | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1048310MM 1ML
Encompass_Preferred
SCH-1473759 hydrochloride is a small-molecule aurora kinase inhibitor with potent activity against aurora A (IC50 4 nM) and aurora B (IC50 13 nM). It shows antiproliferative activity across multiple tumor cell lines and has demonstrated tumor growth inhibition in xenograft models. Available as solid material and as a ready-to-use 10 mM solution in DMSO for research applications.
  • Potent inhibition of aurora A and aurora B (IC50 4 nM and 13 nM).
  • Demonstrated antiproliferative activity in diverse tumor cell lines.
  • Shown in vivo tumor growth inhibition in xenograft models.
  • Available as solid and as a convenient DMSO solution for experiments.
  • High purity suitable for research applications.
  • Documented solubility and formulation protocols for in vitro and in vivo use.
Catalog No. 50-002-43219