Learn More
Cayman Chemical SCH 23390hydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1563110 MG
Encompass_Preferred
A selective antagonist of the dopamine D1-like receptor subtypes D1 and D5 (Kis = 0.2 and 0.3 nM, respectively); useful for studying the role of the dopamine system in normal brain function and neurological disorders
Catalog No. 50-297-2452