Learn More
TARGETMOL CHEMICALS INC SCH-336 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T247715MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SCH-336 is a CB2 receptor agonist (Ki-1.8 nM EC50-2 nM) with effective selective and oral activity. SCH-336 is also bioactive against CB1 receptor and is 100 times more selective to CB2 receptor than CB1. SCH-336 reduced the binding of guanosine 5' -3-o -(sulfur) triphosphate to the membrane containing hCB inhibited BaF3/CB2 cell migration inhibited leukocyte migration in mouse delayed hypersensitivity models and inhibited antigen-induced pulmonary eosinophilia in mouse allergy models. purity: 95%
Catalog No. 50-258-0783