Learn More
Selleck Chemical LLC SCH58261
SDP
Supplier:  Selleck Chemical LLC S81041G
Encompass_Preferred
SCH 58261 is a potent and selective A2a adenosine receptor antagonist with Ki of 2 3 nM and 2 nM for rat A2a and bovine A2a respectively
Catalog No. 50-000-00230