Learn More
Medchemexpress LLC HY-50846 10mg Medchemexpress, SCH772984 CAS:942183-80-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY5084610MG
Encompass_Preferred
Medchemexpress, HY-50846 10mg SCH772984 CAS:942183-80-4 SCH772984 is a highly selective and ATP-competitive ERK inhibitor, with IC50s of 4 and 1 nM for ERK1 and ERK2, respectively. SCH772984 has antitumor activity in MAPK inhibitor-naïve and MAPK inhibitor-resistant cells containing BRAF or RAS mutations. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9337