Learn More
Apexbio Technology LLC SCH772984 HCl, 25mg. Cas: 942183-80-4 (free base) MFCD: NA. ERK1/2 inhibitor
SDP
Supplier:  Apexbio Technology LLC B586625
Encompass_Preferred
SCH772984 HCl is a specific inhibitor of extracellular signal-regulated kinase 1/2 (ERK1/2), with IC50 values of 4 nM and 1 nM, respectively. ERK1/2 is a mitogen-activated protein kinase (MAPK) which is involved in the development of various cancers, especially the ones with BRAF or RAS mutations. Phospho-ERK reactivation in cancer cells is associated with resistance to BRAF and MEK inhibitors. Other sizes are available. Please inqury us for quote.
Catalog No. NC1920465