Learn More
Medchemexpress LLC HY-15532 5mg , SCH900776 CAS:891494-63-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY155325MG
Encompass_Preferred
HY-15532 5mg SCH900776 CAS: 891494-63-6 SCH900776 (MK-8776) is a potent, selective and orally bioavailable inhibitor of checkpoint kinase1 (Chk1) with an IC50 of 3 nM. SCH900776 shows 50- and 500-fold selectivity over CDK2 and Chk2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9752