Learn More
Bioworld Schenk and Hildebrandt Vitamin Concentration (Syringe Vials), 100 mL
Supplier:  Bioworld 306301131
Encompass_Preferred
Schenk & Hildebrandt Vitamin ConcentrationContains Vitamins as described by Schenk & Hildebrandt (1972)Package includes 10 prepared, filtered, and sterilized ampules of 10 mL of 100X vitamin stock solution Use 10 mL of solution to prepare 1 liter of mediumContents (1X): myo Inositol: 1000.0 mg/LNicotinic acid: 5.0 mg/LPyridoxine HCl: 0.5 mg/LThiamine HCl: 5.0 mg/L
Catalog No. 50-254-783