Learn More
Selleck Chemical LLC Schisandrin A S3822-5mg
SDP
Supplier:  Selleck Chemical LLC S38225MG
Encompass_Preferred
Schisandrin A (Sch A Deoxyschizandrin Wuweizisu A) is an active component of Schisandrae Fructus with liver-protective antitumor and antioxidant activities It is an agonist of the adiponectin receptor 2 (AdipoR2) with the IC50 value of 3 5 M
Catalog No. 50-310-9642