Learn More
Selleck Chemical LLC Schisantherin A S3880-10mg
SDP
Supplier:  Selleck Chemical LLC S388010MG
Encompass_Preferred
Schisantherin A (Gomisin C Schisanwilsonin H Arisanschinin K) is a dibenzocyclooctadiene that exhibits anti-tussive sedative anti-inflammatory anti-osteoporotic neuroprotective cognition enhancing and cardioprotective activities Schisantherin A inhibits p65-NF- B translocation into the nucleus by I B degradation
Catalog No. 50-311-1510