Learn More
Cayman Chemical SchIsndrIn A 50mg
SDP
Supplier:  Cayman Chemical 1984950 MG
Encompass_Preferred
A lignan with diverse biological activities; binds to adiponectin receptor 2 (IC50 = 3.2 µM); reduces LPS-induced production of NO, IL-6, and TNF-α and apoptosis in primary mouse microglial cells at 10, 20, and 50 µM; reverses P-glycoprotein-mediated doxorubicin resistance in MCF-7/dox cells at 20 µM; reverses Aβ42-induced short-term and spatial memory deficits, as well as decreases in cerebral SOD and GPX activities in a mouse model of Alzheimer's disease at 4, 12, and 36 mg/kg
Catalog No. 50-298-2411