Learn More
Cayman Chemical SchIsndrIn C 10mg
SDP
Supplier:  Cayman Chemical 2747510 MG
Encompass_Preferred
A lignan with diverse biological activities; decreases viability of U937 cells induces cell cycle arrest at the G1 phase; decreases hydrogen peroxide-induced cell death and production of ROS in C2C12 skeletal muscle cells from 5-20 μM; decreases levels of MMP-2, MMP-9, COX-2, VCAM-1, IL-1β, and TNF-α in hydrogen peroxide-stimulated C2C12 cells; decreases lipoteichoic acid-induced production of NO, PGE2, TNF-α, IL-1β, and IL-6 in mouse primary microglia; decreases serum ALT and AST activity, increases hepatic mitochondrial and total GSH levels, and reduces liver injury in a mouse model of acetaminophen-induced liver injury
Catalog No. 50-294-1173